Ready-to-Load™ DNA Electrophoresis, Genetic Flightplan
Proveedor: EDVOTEK
921
EDVO921EA 109 EUR
EDVO921
Ready-to-Load™ DNA Electrophoresis, Genetic Flightplan
Sistemas de electroforesis Sistemas horizontales de electroforesis en gel
The objective of this experiment is to investigate the genetic basis of beak size variation in Galapagos finches before and after a drought event using Restriction Fragment Length Polymorphism (RFLP) analysis.
  • Introduces PCR-RFLP analysis to analyse specific nucleotide sequences
  • Understand how electrophoresis separates differently-sized molecules
  • Perform agarose gel electrophoresis
  • Complete in 45 minutes
By analysing DNA samples collected from finches with different beak sizes, students determine the association between specific genetic variations andMore Product Information