Experimentation kits, analysis and comparision of mammalian cell types
Proveedor: EDVOTEK
986
EDVO986EA 149 EUR
EDVO986
Experimentation kits, analysis and comparision of mammalian cell types
Materiales educativos Materiales educativos de biología Physiology and Health Learning Activities
Observe the differences between mammalian cell types and how these cells funciton. Cells are fixed on microscope slides and students stain the cells on the slide to view morphological characteristics of the cell types. These cells are very safe for classroom use.
  • For 6 lab groups
  • Complete in 35 minutes
  • Requires a microscope with 400x magnification
Información de suministro: Kit includes instructions, multispot slides (4 cell types each), eosin and methylene blue stain, mounting medium, cover slips, transfer pipettes, and immersion troughs.