Micropipetting Basics
Proveedor: EDVOTEK
S-44
EDVOS-44EA 60.5 EUR
EDVOS-44
Micropipetting Basics
Materiales educativos Materiales educativos de seguridad en laboratorio
Kit for teaching students how to use a micropipette with ease and accuracy using multi-coloured dyes.
  • 10 lab groups
  • Complete in 45 minutes
Also required: 5 to 50 µl adjustable or 10 µl fixed micropipette and tips.
Información de suministro: Kit includes instructions, various coloured dye samples and a pipette card.