Centering Ring, KF25SVCR
Proveedor: EDWARDS HIGH VACUUM METTLER
KF25SVCR
EDWAKF25SVCREA 19.8 EUR
EDWAKF25SVCR
Centering Ring, KF25SVCR
Juntas Anillos de Sellado
Used for vacuum hose connection to rotary pump.
  • Stainless steel with a black o-ring
  • Compatiable with all Edwards and Leybold pump options