Dicofol

Proveedor: EHRENSTORFER

C12570000
EHERC12570000EA 165 EUR
EHERC12570000
Dicofol
Dicofol
Formula: C₁₄H₉Cl₅O
MW: 370.49 g/mol
Núm. CAS: 115-32-2
EINECS: 204-082-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR