JALAS® 2562 COOL ESD Occupational Sandals
Proveedor: EJENDALS
2562-35 2562-36 2562-37 2562-38 2562-39 2562-40 2562-41 2562-42 2562-43 2562-44 2562-46 2562-47 2562-48 2562-45
EJEN2562-35EA 98.23 EUR
EJEN2562-35 EJEN2562-36 EJEN2562-37 EJEN2562-38 EJEN2562-39 EJEN2562-40 EJEN2562-41 EJEN2562-42 EJEN2562-43 EJEN2562-44 EJEN2562-46 EJEN2562-47 EJEN2562-48 EJEN2562-45
JALAS® 2562 COOL ESD Occupational Sandals
Zapatos y botas
Regular fitting, upper material is made of nubuck leather, hook and loop fastening. It prevents risk of antistatic and slipping.
  • Oil-resistant outsole
  • Light with excellent grip
  • Perfect fit and extra comfortable
  • Very breathable
  • Ergothan shock absorption system
Mainly used for indoors, office environments, dry environments, clean environments and primary used in electrical installation, electronics work, laboratory work, cleaning, hotel- and restaurant work, healthcare, care work, service work, office work, shop work.
Certificaciones: EN ISO 20347:201More Product Information