ESD shoes, lace-up, JALAS® Green Line 6468 EKO
Proveedor: EJENDALS
6468-36 6468-37 6468-38 6468-39 6468-40 6468-41 6468-42 6468-43 6468-44 6468-45 6468-46 6468-47
EJEN6468-36EA 135.56 EUR
EJEN6468-36 EJEN6468-37 EJEN6468-38 EJEN6468-39 EJEN6468-40 EJEN6468-41 EJEN6468-42 EJEN6468-43 EJEN6468-44 EJEN6468-45 EJEN6468-46 EJEN6468-47
ESD shoes, lace-up, JALAS® Green Line 6468 EKO
Zapatos y botas
JALAS® Green Line is the first collection of safety footwear to be given the EU Ecolabel. The Ecolabel certificate is only awarded to products that meet the strict limit values specified for energy consumption and environmental friendliness and do not contain chemicals that are harmful to people.
  • Water repellent
  • Oil-resistant outsole
  • Ergothan Shock Absorption system
  • Anti-static properties
  • ProNose toe reinforcement
Metal-free construction with nail protection in plasma-treated composite (PTC) textile, toe cap in composite, and the FX2 Supreme insoleMore Product Information