Reverse osmosis water purification system, PURELAB® Chorus 3
Proveedor: ELGA LabWater
RO310BPM1 RO310XXM1 RO320BPM1 RO320XXM1 RO330BPM1 RO330XXM1
ELGARO310BPM1EA 0 EUR
ELGARO310BPM1 ELGARO310XXM1 ELGARO320BPM1 ELGARO320XXM1 ELGARO330BPM1 ELGARO330XXM1
Reverse osmosis water purification system, PURELAB® Chorus 3
Purificación de agua Sistemas de agua pura y ultra pura
PURELAB Chorus 3 provides Type III (primary RO grade) water on demand.
  • Easy to install, operate and maintain with a clear indication of water purity
  • Economical: Optional CO₂ removal from the purified water (post RO) increasing the life of downstream consumables
  • Option to reduce water consumption for low hardness feed waters
  • Configuration: Ability to configure multiple systems to increase the flow rate
  • Auto Rinse: Maintains purity of water during periods of low use
Typical applications include: Glassware rinsing, heating baths, autoclave filling,More Product Information