Spurr low viscosity embedding kit, Sigma-Aldrich®

Proveedor: SIGMA ALDRICH MICROSCOPY

EM0300-1KT
EM0300-1KTEA 273 EUR
EM0300-1KT
Spurr low viscosity embedding kit, Sigma-Aldrich®
Tintes Reactivos de microscopía y tinción

Spurr resin is a hydrophobic resin. It is less viscous than epon mixtures and is easier to dispense. The low viscosity of the resin contributes to its penetration in tissue, mineral and dense structures. A low viscosity of 60 cps also allows easy penetration into a variety of difficult materials.


  • Quality Level 200


Exceptional penetration for tissue, mineral, and dense structures. A low viscosity of 60 cps allows easy penetration into a variety of difficult materials. Spurr low viscosity embedding kit has been used to embed histological samples like tissues, organs and embryos.[2][3][4] It may be used to prepare mineral specimens for polishing. The low viscosity of the Spurr formulation allows rapid infiltration into tissues, minerals, and other dense structures.


Kit contains:


Nonenylsuccinic anhydride 450 g
D.E.R. 736 250 g
Dimethylaminoethanol 100 g
ERL 4221 250 g

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR