Exonuclease I
Proveedor: Thermo Fisher Scientific
EN0581 EN0582
EN0581EA 110 EUR
EN0581 EN0582
Exonuclease I
Enzimas
Exonuclease I (ExoI) degrades single-stranded DNA in a 3' to 5'direction. It releases deoxyribonucleoside 5'-monophosphates in a stepwise manner and leaves 5'-terminal dinucleotides intact.
Used for primer removal from PCR mixtures, prior to PCR product sequencing, for one-tube 'megaprimer' PCR mutagenesis, removal of single-stranded DNA containing a 3'-hydroxyl terminus from nucleic acid mixtures, assay for the presence of single-stranded DNA with a 3'-hydroxyl terminus.
Precaución: For research use only. Not for use in diagnostic procedures.
Order Now
ESPECIFICACIONES
Nombre de la enzima Exonuclease I Exonuclease I
Environmentally Preferable
Fuente
Especies
Concentración 20 U/µl 20 U/µl