Anti-BECN1 Rabbit Polyclonal Antibody

Proveedor: ENZO LIFE SCIENCES
ADI905721100
ENZOADI905721100EA 510 EUR
ENZOADI905721100
Anti-BECN1 Rabbit Polyclonal Antibody
Anticuerpos
Beclin-1 is a coiled-coil Bcl-2-interacting protein that functions in autophagy, the process of bulk degradation of cellular proteins. Beclin-1 is homologous to the yeast autophagy gene apg6, and is expressed at reduced levels in human breast carcinoma, suggesting that defects in autophagy proteins may contribute to the development or progression of tumors. Bcl-2 can inhibit Beclin-1-dependent autophagy in yeast and mammalian cells, suggesting that Bcl-2 functions as both an anti-autophagic and anti-apoptotic protein.

  • For Immunohistochemistry, Western Blot

Recognizes human, mouseMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno BECN1
Environmentally Preferable
Nombre del antígeno Beclin 1
Sinónimos de antígenos Coiled-coil myosin-like BCL2-interacting protein|APG 6|VPS30|Beclin1 autophagy|ATG6|CTIP-1 COUP-TF interacting protein 1|Protein GT197
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Reactividad
Huésped Rabbit
Isotipo
Western blot Yes
InmunoQuímica Yes
Forma Liquid. In PBS containing 0,02% sodium azide.
Purification Peptide affinity purified

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR