IHC reagent (mouse/rabbit-HRP), POLYVIEW®
Proveedor: ENZO LIFE SCIENCES
ADI9501130100
ENZOADI9501130100EA 550 EUR
ENZOADI9501130100
IHC reagent (mouse/rabbit-HRP), POLYVIEW®
Ensayos Ensayos inmunológicos
Immunohistochemistry (mouse/rabbit-HRP) detection reagent is a non-biotin one-step detection reagent.
  • Biotin-free nanopolymer detection circumvents endogenous biotin background
  • High intensity colour development, which delivers sharp and crisp staining
  • Used with blood smears, cytosmears, and cell preparations
Suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and frozen sections.