Lysates, HeLa (human)
Proveedor: ENZO LIFE SCIENCES
ADILYCHL100F ADILYCHL101F ADILYCHL102F
ENZOADILYCHL100FEA 165 EUR
ENZOADILYCHL100F ENZOADILYCHL101F ENZOADILYCHL102F
Lysates, HeLa (human)
Células
The HeLa cell line derived from a human donor's epithelioid carcinoma tissue (cervix).
Dilution range: 20 µg/lane (Western blotting)
Liquid lysates in SDS-PAGE sample buffer.
Heat shock-treated HeLa cells with no recovery time can be a useful tool in studying the regulators of immediate-early gene expression during heat shock stress response.