Anti-HSP90B1 Rat Monoclonal Antibody [clone: 9G10]

Proveedor: ENZO LIFE SCIENCES
ADISPA850PED ADISPA850PEF
ENZOADISPA850PEDEA 305 EUR
ENZOADISPA850PED ENZOADISPA850PEF
Anti-HSP90B1 Rat Monoclonal Antibody [clone: 9G10]
Anticuerpos
Type: Primary
Antigen: HSP90B1
Clonality: Monoclonal
Clone: 9G10
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Pig, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR