Nodularin, linear
Proveedor: ENZO LIFE SCIENCES
ALX350182C100
ENZOALX350182C100EA 283 EUR
ENZOALX350182C100
Nodularin, linear
Nodularin
Precursor of the highly toxic cyclic pentapeptide, nodularin.
Isolated from Nodularia spumigena.
Linear nodularin has been demonstrated to be a biosynthetic precursor of the highly toxic cyclic pentapeptide, nodularin. It is an intermediate of biodegradation by natural populations of bacteria in aquatic samples.
Formula: C₄₁H₆₀N₈O₁₀
Núm. CAS: 118399-22-7