S-Nitrosoglutathione (GSNO) ≥98% (by TLC)

Proveedor: ENZO LIFE SCIENCES

Sinónimos: (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-(nitrosothio)-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, GSNO, Nitrosoglutathione, S-Nitroso-L-Glutathione

ALX420002M025 ALX420002M100
ENZOALX420002M025EA 136 EUR
ENZOALX420002M025 ENZOALX420002M100
S-Nitrosoglutathione (GSNO) ≥98% (by TLC)
S-Nitrosoglutathione (GSNO)

Nitric oxide donor


NO donor which spontaneously releases NO under physiological conditions. Half-life of 159 hours under physiological conditions. Selectively inhibits platelet activation and displays cardioprotective effects. Inhibits NF-kappaB activation. Inhibits endothelial cell proliferation and induces apoptosis in T cells.

Formula: C₁₀H₁₆N₄O₇S
MW: 336.319 g/mol
Número MDL: MFCD00214341
Núm. CAS: 57564-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR