Calcium pantothenate ≥98% (by HPLC)

Proveedor: ENZO LIFE SCIENCES

Sinónimos: Calcium D-Pantothenate, D-Pantothenic acid calcium salt, D-Calcium pantothenate, Calcium (R)-3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoate, Vitamin B5 calcium salt, Calcium D-(+)-pantothenate

ALX460003G010
ENZOALX460003G010EA 50.6 EUR
ENZOALX460003G010
Calcium pantothenate ≥98% (by HPLC)
Calcium pantothenate

Precursor for coenzyme A biosynthesis.


Soluble in water. Also soluble in glycerol; slightly soluble in 100% ethanol or methanol; insoluble in acetone or ether.

Formula: C₁₈H₃₂CaN₂O₁₀
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00002766
Núm. CAS: 137-08-6
EINECS: 205-278-9
Índice Merck: 14,07015

Order Now

Resultados test especificaciones

Purity (HPLC) ≥98%
Identity Determined by IR
Appearance White to off-white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR