Human Recombinant CD137:Fc
Proveedor: ENZO LIFE SCIENCES
ALX522031C050
ENZOALX522031C050EA 587 EUR
ENZOALX522031C050
Human Recombinant CD137:Fc
Proteínas y Péptidos
Produced in HEK 293 cells. The cysteine-rich region of human CD137 (4-1BB) (aa 1-186) is fused to the Fc portion of human IgG1.