Human Recombinant EDA-A1 (soluble) (from E. coli)
Proveedor: ENZO LIFE SCIENCES
ALX522038C010
ENZOALX522038C010EA 590 EUR
ENZOALX522038C010
Human Recombinant EDA-A1 (soluble) (from E. coli)
Proteínas y Péptidos
Produced in E. coli. The extracellular domain of human EDA-A1 (ectodysplasin-A1) (aa 245-391) is fused at the N-terminus to a linker peptide (9 aa) and a FLAG®-tag.