Mouse recombinant CD40L, soluble
Proveedor: ENZO LIFE SCIENCES
ALX850075KI01
ENZOALX850075KI01EA 917 EUR
ENZOALX850075KI01
Mouse recombinant CD40L, soluble
Proteínas y Péptidos
ELISAFunctional Application: Stimulates the proliferation of mouse B cells and dendritic cells. The activity of CD40L increases 1'000-fold (stimulation in the ng/ml range) in the presence of the cross-linking enhancer (Prod. No. ALX-804-034).
Información para pedidos: Contents: 10 µg of CD40L, Soluble (mouse) (recombinant) (Prod. No. ALX-522-070) and 2×50 µg of Enhancer for Ligands (Prod. No. ALX-804-034) which increases the biological activity of CD40L at least 1'000-fold.