CD14, soluble (human) ELISA Kit
Proveedor: ENZO LIFE SCIENCES
ALX850302KI01
ENZOALX850302KI01EA 1030 EUR
ENZOALX850302KI01
CD14, soluble (human) ELISA Kit
Ensayos ELISAs
For the quantitative determination of soluble human CD14 in serum, plasma and cell culture medium.
Información para pedidos: Contains 1 precoated ELISA module1 vial detecting antibody (POD-labelled monoclonal antibody to human CD14) (ready-to-use)1 vial human CD14 standard (recombinant human CD14, lyophilized)1 vial reference serum (2.7 ± 0.5, lyophilized)2 tablets PBS1 vial dilution buffer1 vial Tween 201 vial stopping solution (ready-to-use)1 vial substrate solution (ready-to-use).