all cis-8,11,14-Eicosatrienoic acid ≥99% (por GC), clear liquid, grado ultrapuro

Proveedor: ENZO LIFE SCIENCES

BMLFA0090100 BMLFA0091000
ENZOBMLFA0090100EA 165 EUR
ENZOBMLFA0090100 ENZOBMLFA0091000
all cis-8,11,14-Eicosatrienoic acid ≥99% (por GC), clear liquid, grado ultrapuro
all cis-8,11,14-Eicosatrienoic acid

Beneficial effects in cardiovascular diseases and inflammation.


Isolated from borage oil.


Soluble in ethanol, DMSO or acetone.

Formula: C₂₀H₃₄O₂
Número MDL: MFCD00065721
Núm. CAS: 1783-84-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f9a9c72155f78b6330bdd5f6472c1b75ab3e21e&vid=a98ed0c828f17bcd7fe4480c35b8ba9d654d58d2&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Purity ≥99% (GLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR