12(R)-Hydroxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (HETE) ≥99%

Proveedor: ENZO LIFE SCIENCES

Sinónimos: 12(R)-HETE

BMLH1120050
ENZOBMLH1120050EA 324 EUR
ENZOBMLH1120050
12(R)-Hydroxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (HETE) ≥99%
12(R)-Hydroxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (HETE)

Biosynthesis in psoriatic skin and corneal epithelial cells via cytochrome P-450. More potent than 12(S)-HETE in inducing human leukocyte chemokinesis and aggregation. Proinflammatory effects in human skin. Inhibits renal, cardiac and corneal Na+,K+-ATPase. Reduces intraocular pressure.


  • ATPase inhibitor


Biosynthesis in psoriatic skin and corneal epithelial cells via cytochrome P-450. More potent than 12(S)-HETE in inducing human leukocyte chemokinesis and aggregation. Proinflammatory effects in human skin. Inhibits renal, cardiac and corneal Na+,K+-ATPase. Reduces intraocular pressure.

Formula: C₂₀H₃₂O₃
MW: 320,5 g/mol
Temperatura de almacenaje: Ultracongelador (–80°C)
Núm. CAS: 82337-46-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e78cd8129c69f840beec88ed472c856174cc4eee&vid=9d8f88eb0ea2dc5774ae1143a49ed6f3790acb78&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR