AAF-CMK
Proveedor: ENZO LIFE SCIENCES
BMLPI1230025
ENZOBMLPI1230025EA 337 EUR
ENZOBMLPI1230025
AAF-CMK
Proteínas y Péptidos
Tripeptidyl peptidase inhibitor
A serine protease inhibitor which irreversibly inhibits tripeptidyl peptidase II (TPPII), a giant protease which may substitute for some proteasome functions, and reversibly inhibits TPPI. Shows only marginal inhibition of the chymotrypsin-like activity of the proteasome.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=069b56833e3d41c686de55282a26551af326f62e&vid=46444ce5010aef4aa9804f7722f623ddbb0b4e4c&prid=web&clid=SAPDAM