Anti-Proteasome 19S Rpn10/S5a subunit Rabbit Polyclonal Antibody

Proveedor: ENZO LIFE SCIENCES
BMLPW10450025 BMLPW10450100
ENZOBMLPW10450025EA 161 EUR
ENZOBMLPW10450025 ENZOBMLPW10450100
Anti-Proteasome 19S Rpn10/S5a subunit Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Proteasome 19S Rpn10/S5a subunit
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno Proteasome 19S Rpn10/S5a
Environmentally Preferable
Nombre del antígeno Proteasome 19S Rpn10/S5a Subunit
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human
Huésped Rabbit
Isotipo
Western blot Yes
Inmunógeno Full-length recombinant GST-tagged human S5a (Accession number: P55036)
Forma Liquid
Purification Purified by immunodepletion of GST-reactive antibodies followed by Protein A purification
Storage buffer PBS containing 0,01% sodium azide
Temperatura de almacenamiento –20 °C
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR