Kifunensine ≥98% (by TLC)

Proveedor: ENZO LIFE SCIENCES

BMLS1140001 BMLS1140005
ENZOBMLS1140001EA 154 EUR
ENZOBMLS1140001 ENZOBMLS1140005
Kifunensine ≥98% (by TLC)
Kifunensine

Inhibits α-mannosidase. Inhibits asparagine-linked oligosaccharide processing. Immunomodulator.


  • Mannosidase inhibitor


White to off-white solid.


Soluble in water (25mg/ml).

Formula: C₈H₁₂N₂O₆
Temperatura de almacenaje: Congelador
Núm. CAS: 109944-15-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91d671159b983a2b9b447ff141e04a99a4a9566d&vid=7f485c19a1e14251b738b9f4a42d53cc3a28b823&prid=web&clid=SAPDAM

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR