Anti-GCG Mouse Monoclonal Antibody [clone: 8]

Proveedor: ENZO LIFE SCIENCES
BPDABS058081
ENZOBPDABS058081EA 4130 EUR
ENZOBPDABS058081
Anti-GCG Mouse Monoclonal Antibody [clone: 8]
Anticuerpos
Type: Primary
Antigen: Glucagon
Clonality: Monoclonal
Clone: 8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat, Pig
New Product
Order Now

ESPECIFICACIONES

Simbolo antígeno GCG
Nombre del antígeno Glucagon
Environmentally Preferable
Sinónimos de antígenos GCG|GLP2|GRPP|Glicentin-related Polypeptide|Glucagon-like Peptide 2|Glucagon-like Peptide 1|GLP1
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Clon 8
Reactividad Pig,Human,Rat
Huésped Mouse
Isotipo IgG1 kappa
ELISA Yes
Epítopo N-Terminal
Inmunógeno Synthetic N-terminal fragment corresponding to human glucagon coupled to carrier
Forma Liquid
Purification Purified by Protein A affinity chromatography
Storage buffer 0,01 M phosphate buffer, pH 7,4, containing 0,5 M sodium chloride and 15 mM sodium azide
Temperatura de almacenamiento 4 °C
Shipping temperature Blue ice not frozen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR