Antifade mounting medium
Proveedor: ENZO LIFE SCIENCES
ENZ53002M010 ENZ53003M010
ENZOENZ53002M010EA 165 EUR
ENZOENZ53002M010 ENZOENZ53003M010
Antifade mounting medium
Tintes Reactivos de microscopía y tinción
Antifade mounting medium (hardset) is an antifade reagent that causes little or no quenching. Use of the mounting medium significantly extends the photobleaching half-life of the fluorescent stain.
  • Inhibits photobleaching of fluorescent dyes and fluorescent proteins
  • Hardening formulation
Recommended for use on fixed cells and tissues that have been stained with fluorescent stains. Cells can be viewed immediately, or if long term storage is desired, the reagent can be allowed to harden overnight.
New Product