Anti-CD40 Mouse Monoclonal Antibody (Biotin) [clone: G28-5]

Proveedor: ENZO LIFE SCIENCES
ENZABS1290100
ENZOENZABS1290100EA 634 EUR
ENZOENZABS1290100
Anti-CD40 Mouse Monoclonal Antibody (Biotin) [clone: G28-5]
Anticuerpos
Recommended Applications: Flow Cytometry, IHC (PS)

Species reactivity: Human

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: G28-5
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR