Human growth hormone ELISA kit
Proveedor: ENZO LIFE SCIENCES
ENZKIT1480001
ENZOENZKIT1480001EA 630 EUR
ENZOENZKIT1480001
Human growth hormone ELISA kit
Ensayos ELISAs
Ultra-sensitive human growth hormone ELISA kit enabling detection of HGH in serum, plasma, saliva and tissue culture media in just 2,5 hours.
  • Ultra-sensitive measurement of HGH, detecting as little as 0,93 pg/ml
  • Negligible cross reactivity with similar proteins (prolactin, human placental lactogen)
  • High throughput format with results in 2,5 hours for up to 36 samples in duplicate
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of growth hormone in serum, plasma, saliva and tissue culture media. The human growth hormone ELISA kit is a colorimetric immunometric immunoassay kit with results in 2,5 hours.
Información para pedidos: Contains Microtiter plate, Assay Buffer 13, Standard, Wash Buffer Concentrate, Antibody, Antibody Diluent, Conjugate, Substrate,  and Stop Solution.