Jars, sampling, wide mouth, with screw cap
Proveedor: EP SCIENTIFIC
120-0125 321-2000 320-0125 320-0250 320-0500 321-0125 321-0250 321-0500 321-1000 121-4000 121-0250 120-0060 121-1000 120-0250 120-2000 121-0125 221-2000 220-0250 220-0500 220-2000 221-0125 221-0250 220-0060 221-1000 221-4000 221-0500 220-0125 320-0060 320-2000 321-4000 V320-0125 V220-0125 V320-0500 V321-1000 V221-1000
EPSC120-0125EA 175 EUR
EPSC120-0125 EPSC321-2000 EPSC320-0125 EPSC320-0250 EPSC320-0500 EPSC321-0125 EPSC321-0250 EPSC321-0500 EPSC321-1000 EPSC121-4000 EPSC121-0250 EPSC120-0060 EPSC121-1000 EPSC120-0250 EPSC120-2000 EPSC121-0125 EPSC221-2000 EPSC220-0250 EPSC220-0500 EPSC220-2000 EPSC221-0125 EPSC221-0250 EPSC220-0060 EPSC221-1000 EPSC221-4000 EPSC221-0500 EPSC220-0125 EPSC320-0060 EPSC320-2000 EPSC321-4000 EPSCV320-0125 EPSCV220-0125 EPSCV320-0500 EPSCV321-1000 EPSCV221-1000
Jars, sampling, wide mouth, with screw cap
Tarros Tarros para uso general
Jars, clear or amber Type III soda-lime glass, with PTFE-lined white polypropylene closed-top cap, designed for collecting soil, sludge, waste and water samples for a variety of organic and inorganic target analyses.
  • Certified to meet EPA performance-based specifications for semi-volatile organics, pesticides, PCBs and metals analyses
  • Processed the same way as the certified jars but without certification documentation
  • Unprocessed and ready for your own cleaning procedure
  • Closures assembled on bottles
  • Available as short or tall profile
Información deMore Product Information