Jugs, with handle
Proveedor: EP SCIENTIFIC
115-4000 315-4000 315-2000
EPSC115-4000EA 95.4 EUR
EPSC115-4000 EPSC315-4000 EPSC315-2000
Jugs, with handle
Frascos Garrafones
HDPE, natural, with white LDPE foam-lined PP screw closure.
  • Good chemical resistance
  • Good temperature resistance
The narrow mouth and sturdy handle make carrying and pouring higher volume samples easy. These are recommended for collecting water samples to be analyzed for inorganic trace analytes. Unprocessed or certified available.
Meet or exceed EPA performance-based standards for metals, cyanide and fluoride.
Order Now
ESPECIFICACIONES
Environmentally Preferable Low Manufacturing Impact
Certificación ambiental NAR Certificate of Retirement