Restriction enzymes, NotI, 50 U/µl, Fermentas
Proveedor: Thermo Fisher Scientific
ER0593
ER0593EA 283 EUR
ER0593
Restriction enzymes, NotI, 50 U/µl, Fermentas
Reactivos de ácidos nucleicos Restriction Enzymes
NotI restriction enzyme recognises GC^GGCCGC sites and cuts best at 37 °C in O buffer.
  • Superior quality, stringent quality control and industry leading manufacturing process
  • Convenient colour-coded five buffer system
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Applications: Molecular cloning, restriction site mapping, genotyping, southern blotting, restriction fragment length polymorphism (RFLP), SNP.