Floor signs, DuraStripe®, X-Treme, corner
Proveedor: ERGOMAT
DSXCR4B DSXTR4B DSXCR4Y DSXTR4Y DSXSTEPY 4070058XTCG 4070058XF9BL 4070058XF9Y
ERGODSXCR4BEA 99.9 EUR
ERGODSXCR4B ERGODSXTR4B ERGODSXCR4Y ERGODSXTR4Y ERGODSXSTEPY ERGO4070058XTCG ERGO4070058XF9BL ERGO4070058XF9Y
Floor signs, DuraStripe®, X-Treme, corner
Señales
DuraStripe® is the easiest, fastest and least expensive floor marking system for industrial floors. The durability and vast size of the DuraStripe® program makes it the only total floor marking solution available on the market today. Economical than painting the floors.
  • Ideal for smooth or newer floors but can be used on old floors as well
  • Simple and easy peel-&-stick installation
  • Easy to remove for a quick reconfiguration and hence a great Lean Manufacturing tool
  • No dry time, cure time, fumes or messy clean up to limit productivity
  • Available in 11