Cleanroom anti-fatigue mats, Infinity Bubbles
Proveedor: ERGOMAT
IN0304-W IN0420-W IN0420-B IN0412-W IN0412-B IN0406-W IN0406-B IN0320-W IN0320-B IN0306-B IN0304-B IN0220-W IN0220-B IN0209-W IN0209-B IN0206-W IN0206-B IN0203-W IN0203-B IN0306-W IN0409-W IN0404-W IN0409-B IN0303-B IN0303-W IN0404-B IN0212-B IN0212-W IN0312-B IN0312-W IN0309-W IN0309-B IN0204-B IN0204-W
115-0931EA 327 EUR
115-0931 115-0936 115-0946 115-0935 115-0945 115-0934 115-0944 115-0933 115-0943 115-0942 115-0941 115-0930 115-0940 115-0929 115-0939 115-0928 115-0938 115-0927 115-0937 115-0932 ERGOIN0409-W ERGOIN0404-W ERGOIN0409-B ERGOIN0303-B ERGOIN0303-W ERGOIN0404-B ERGOIN0212-B ERGOIN0212-W ERGOIN0312-B ERGOIN0312-W ERGOIN0309-W ERGOIN0309-B ERGOIN0204-B ERGOIN0204-W
Cleanroom anti-fatigue mats, Infinity Bubbles
Alfombrillas Alfombrillas antifatiga
The Infinity Bubble mat has the comfort of a polyurethane mat and easy clean properties due to its closed-cell and non-porous top surface. It is UV and chemical resistant (except for organic solvents).
  • Upper side with bubbles
  • Moulded bevelled edges prevent curling, eliminates trip hazards
  • Virgin raw material guarantee durability and ergonomic benefits
  • Chemical and UV exposure will not affect the state of the top of the mat
  • Closed cell and non-porous surface allows easy wash down and cleaning
  • Solid mat throughout, no hollow bubbles that collapse
  • Anti-static,