Ductless filtering fume hoods, Captair® Smart 481
Proveedor: ERLAB CAPTAIRE
AA626A00011A21
ERLAAA626A00011A21EA 6200 EUR
ERLAAA626A00011A21
Ductless filtering fume hoods, Captair® Smart 481
Vitrinas de gases y recintos Vitrinas de gases sin extracción externa
The new Captair® Smart ductless filtering hoods are designed with a simple and innovative way of communication. Smart Technology, simple and intuitive communication by light indicates the status of the hood. A simple light pattern will indicate if the airflow is compromised, if there is a fan failure or in case of a filter breakthrough.
  • The Erlab's unique Flex Filtration can manage most laboratory applications
  • New sensors are added for detection of solvents or acids or formalehyde
  • Low power consumption
  • New electronic anemometer monitors face velocity
  • The 6°