Reading magnifiers, visoPOCKET
Proveedor: ESCHENBACH OPTIK
172116 172146 172156
ESCH172116EA 27.9 EUR
ESCH172116 ESCH172146 ESCH172156
Reading magnifiers, visoPOCKET
Lupas de laboratorio
Compact magnifiers with wide field of view and ultra-slim lenses.
  • Diffractive aspheric lens with modern and exquisite design
  • High quality leather available in three colours
  • Leather case protects from scratches
Sutiable for on-the-go use when small print needs magnifying swiftly and easily.