Avantor® ACE® Excel® C18-AR, HPLC/UHPLC Columns, Analytical, 3 µm
Proveedor: Avantor
EXL-119-1002UEA 862 EUR
EXL-119-1002U EXL-119-1003U EXL-119-1046U EXL-119-1202U EXL-119-1203U EXL-119-1246U EXL-119-1502U EXL-119-1503U EXL-119-1546U EXL-119-0246U EXL-119-2502U EXL-119-2503U EXL-119-2546U EXL-119-0346U EXL-119-0503U EXL-119-0546U EXL-119-7502U EXL-119-7503U EXL-119-7546U EXL-119-0502U EXL1192546UMVK
Avantor® ACE® Excel® C18-AR, HPLC/UHPLC Columns, Analytical, 3 µm
Columnas de cromatografía
The Avantor® ACE® Excel® C18-AR phase utilises a specially developed ligand combining a C18 chain with integral phenyl functionality, thus combining the benefits of both C18 and phenyl characteristics into a single phase. This can be used for ‘standard’ C18 column separations, but is additionally recommended for separations that involve compounds containing aromatic functionality.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1,7 to 5 µ