Fast blue BB salt ≥80% (dye content), Sigma-Aldrich®

Proveedor: SIGMA ALDRICH MICROSCOPY
Atención

Sinónimos: 4-Benzoylamino-2,5-diethoxybenzenediazonium chloride hemi(zinc chloride) salt, 4-Amino-2,5-diethoxybenzanilide diazotated zinc double salt, Fast Blue BB Salt hemi(zinc chloride) salt

M-Clarity™ quality level = MQ200

F3378-1G F3378-25G F3378-5G
F3378-1GEA 37.9 EUR
F3378-1G F3378-25G F3378-5G
Fast blue BB salt ≥80% (dye content), Sigma-Aldrich®
Fast blue BB salt

Fast Blue BB Salt hemi (zinc chloride) salt, also known as Cl Azoic diazo 20 (Cl 37175) is a diazonium salt that possesses a lipophilic amine lipophilic with an aromatic system of moderate size. It is commercially available as the zinc chloride double salt. It has been used for alkaline phosphatase staining in cells.

Formula: C₃₄H₃₆Cl₄N₆O₆Zn
MW: 831.89 g/mol
Pto. de fusión: 157 °C
Temperatura de almacenaje: Congelador
Número MDL: MFCD00074765
Núm. CAS: 5486-84-0
EINECS: 226-817-4

Order Now

Resultados test especificaciones

Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Solubility (Turbidity), c = 10 mg/ml; Water Clear to Very Hazy
Solubility (Colour) Yellow to Very Dark Yellow and Brown and Yellow-Brown
Nitrogen ≥8.0%
Dye Content Calculated, Based on Nitrogen Content ≥80%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR