Cleaning mops, PharmaMOP®
Proveedor: FOAMTEC
FT416C FS853 FS858EN FT416D
FOAMFT416CEA 121 EUR
FOAMFT416C FOAMFS853 FOAMFS858EN FOAMFT416D
Cleaning mops, PharmaMOP®
Mopas
PharmaMOP® is designed for all critical cleanroom surfaces. EzGLIDE™ microfiber mop heads aid in protocols that involve cleaning sporicides, bleach, glass and high soil areas such as gowning rooms. It is constructed with a clean-room micro fiber fabric over foam for removing heavy residues from aseptic processing areas. Ideal for SOPs designed to remove bleach and sporicide residues. It is ideal for meeting extra-long contact time requirements to control spores.
  • Ultra-lightweight and applicable for all surfaces
  • Used to clean ceilings, walls and floors with