Giemsa tincione, technical stain_blood, Sigma-Aldrich®

Proveedor: SIGMA ALDRICH MICROSCOPY
Atención

Sinónimos: Tinte de Giemsa

MQ100

G4507-25G G4507-5G
G4507-25GEA 532 EUR
G4507-25G G4507-5G
Giemsa tincione, technical stain_blood, Sigma-Aldrich®
Giemsa tincione

Giemsa stain is a versatile differential stain that is widely used in hematology for blood and bone marrow specimens, clinical cytology specimens, bacteriology, histological biopsies, and tumor samples. Giemsa's staining solution is composed of methylene blue, azure, and eosin. It is a major histological stain due to its unique staining of chromatin, nuclear membranes, and cytoplasmic elements. The stain is frequently used in combination with other dye solutions: May-Grünwald’s solution for Pappenheim (MGG) and Wright-Giemsa.


  • Giemsa stain has been used for the staining of cells and blood films

Pto. de fusión: 300 °C (lit.)
Temperatura de almacenaje: Room Temperature
Número MDL: MFCD00081642
Núm. CAS: 51811-82-6
EINECS: 257-438-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65a3dde4b74ec23e6f527c0ce338d61d470c8d26&vid=c7baef9be683f2300682a16b9432418e0a4311a0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance (Color) Dark to Very Dark Green and Black
Appearance (Form) Powder
Solubility (Color) 4 mg plus 4 ml of Ethanol
Wavelength (1) (UV) 645 - 655 nm
Solvent (UV) Methanol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR