Equipment maintenance and calibration labels, blank
Proveedor: GA
CALA-001-1R CALA-002-1R CALA-010-1R CALA-011-0.5R
GAINCALA-001-1REA 66.8 EUR
GAINCALA-001-1R GAINCALA-002-1R GAINCALA-010-1R GAINCALA-011-0.5R
Equipment maintenance and calibration labels, blank
Etiquetas
These plastic labels can be written on with ball-point pens or permanent markers.
  • Transparent tab will securely laminate the data
  • Protect labels against tampering
Self-laminating, waterproof calibration labels are designed to reliably record the inspection and calibration data on equipment, instruments, and containers used in various industries.