Cryogenic Direct Thermal Labels for Frozen Surfaces
Proveedor: GA
DTU-61NPC1-4WH DTU-7NOTC1-1WH DTU61C3-4BL DTU61C1-4OR DTU227C3-4WH DTU28C1-1WH DTU177C1-075W DTU183C3-4PI DTU158C3-4WH DTU28C3-2WH DTU61C3-4PI DTU61C3-4YE DTU52C1-2WH DTU183C3-4BL DTU183C1-1YE DTU61C1-4PI DTU61C3-4WH DTU183C3-4YE DTU85C3-2WH DTU183C3-4GA DTU52C3-2WH DTU183C3-4WH DTU85C1-1WH DTU284C1-05WH DTU183C1-1GA DTU61C1-4GA DTU183C1-1PI DTU53C3-2WH DTU183C1-1BL DTU61C1-4YE DTU61C1-4BL DTU53C1-2WH DTU61C3-4OR DTU183C1-1WH DTU158C1-1WH DTU7C3-5WH DTU183C1-1OR DTU177C3-3WH DTU227C1-2WH DTU183C3-4OR DTU284C3-1WH DTU61C3-4GA
GAINDTU-61NPC1-4WHEA 165.6 EUR
GAINDTU-61NPC1-4WH GAINDTU-7NOTC1-1WH GAINDTU61C3-4BL GAINDTU61C1-4OR GAINDTU227C3-4WH GAINDTU28C1-1WH GAINDTU177C1-075W GAINDTU183C3-4PI GAINDTU158C3-4WH GAINDTU28C3-2WH GAINDTU61C3-4PI GAINDTU61C3-4YE GAINDTU52C1-2WH GAINDTU183C3-4BL GAINDTU183C1-1YE GAINDTU61C1-4PI GAINDTU61C3-4WH GAINDTU183C3-4YE GAINDTU85C3-2WH GAINDTU183C3-4GA GAINDTU52C3-2WH GAINDTU183C3-4WH GAINDTU85C1-1WH GAINDTU284C1-05WH GAINDTU183C1-1GA GAINDTU61C1-4GA GAINDTU183C1-1PI GAINDTU53C3-2WH GAINDTU183C1-1BL GAINDTU61C1-4YE GAINDTU61C1-4BL GAINDTU53C1-2WH GAINDTU61C3-4OR GAINDTU183C1-1WH GAINDTU158C1-1WH GAINDTU7C3-5WH GAINDTU183C1-1OR GAINDTU177C3-3WH GAINDTU227C1-2WH GAINDTU183C3-4OR GAINDTU284C3-1WH GAINDTU61C3-4GA
Cryogenic Direct Thermal Labels for Frozen Surfaces
Etiquetas
Cryogenic direct thermal labels intended for labeling already frozen surfaces and containers under deep-freeze conditions, as low as (–80 °C).
  • Temperature range: –196 to +70 °C (–321 to +158 °F)
  • Quick labelling of frozen vials and tubes (no thawing required)
  • Withstands exposure to solvents, water baths, and condensation
  • Compatible with most common direct thermal printers
  • Convenient to print, no need for ribbons or other ink sources
These solvent-resistant cryogenic direct thermal printable labels can easily attach to already frozen vials, frozen