Rectangle Permanent Deep Freeze Labels for Thermal Transfer Printers
Proveedor: GA
FJT-10C1-1WH FJT-10C3-2WH FJT-11C1-2WH FJT-11C3-4WH FJT-127C1-1WH FJT-128C1-1WH FJT-128C3-2WH FJT-152C1-1WH FJT162NPC3-2WH FJT-169C1-4WH FJT-169C3-8WH FJT-179C1-1WH FJT-179C3-4WH FJT-18C1-2WH FJT-18C3-2WH FJT-209C1-4WH FJT-209C3-10WH FJT-222C1-2WH FJT-225C1-3WH FJT-238C1-1WH FJT-238C3-2WH FJT-240C1-1WH FJT-240C3-3WH FJT-242C1-1WH FJT-242C3-3WH FJT-24C1-2WH FJT-24C3-2WH FJT-259C1-10WH FJT-27C1-1WH FJT-27C3-3WH FJT-28C1-2WH FJT-28C3-2WH FJT-29C1-1WH FJT-29C3-3WH FJT-36C1-1WH FJT-36C3-1WH FJT-42C1-1WH FJT-42C3-3WH FJT-4C1-1WH FJT-4C3-1WH FJT-504C1-1WH FJT-504C3-3WH FJT506NPC1-4WH FJT-51NPC1-2WH FJT-547C1-1WH FJT-56C1-1WH FJT-56C3-1WH FJT-57C1-1WH FJT-58C1-1WH FJT-58C3-4WH FJT-62C1-0.5WH FJT-62C3-1WH FJT-67C3-1WH FJT-7C1-1WH FJT-7C3-5WH FJT-7SBC1-1WH FJT-85C1-2WH FJT-97C1-1WH FJT-97C3-2WH FJT-98C1-1WH FJT99C1-0.25WH FJT-99C3-0.5WH FJT-9C1-1WH FJT-9C3-1WH FJT-269C1-2WH
GAINFJT-10C1-1WHEA 42 EUR
GAINFJT-10C1-1WH GAINFJT-10C3-2WH GAINFJT-11C1-2WH GAINFJT-11C3-4WH GAINFJT-127C1-1WH GAINFJT-128C1-1WH GAINFJT-128C3-2WH GAINFJT-152C1-1WH GAINFJT162NPC3-2WH GAINFJT-169C1-4WH GAINFJT-169C3-8WH GAINFJT-179C1-1WH GAINFJT-179C3-4WH GAINFJT-18C1-2WH GAINFJT-18C3-2WH GAINFJT-209C1-4WH GAINFJT-209C3-10WH GAINFJT-222C1-2WH GAINFJT-225C1-3WH GAINFJT-238C1-1WH GAINFJT-238C3-2WH GAINFJT-240C1-1WH GAINFJT-240C3-3WH GAINFJT-242C1-1WH GAINFJT-242C3-3WH GAINFJT-24C1-2WH GAINFJT-24C3-2WH GAINFJT-259C1-10WH GAINFJT-27C1-1WH GAINFJT-27C3-3WH GAINFJT-28C1-2WH GAINFJT-28C3-2WH GAINFJT-29C1-1WH GAINFJT-29C3-3WH GAINFJT-36C1-1WH GAINFJT-36C3-1WH GAINFJT-42C1-1WH GAINFJT-42C3-3WH GAINFJT-4C1-1WH GAINFJT-4C3-1WH GAINFJT-504C1-1WH GAINFJT-504C3-3WH GAINFJT506NPC1-4WH GAINFJT-51NPC1-2WH GAINFJT-547C1-1WH GAINFJT-56C1-1WH GAINFJT-56C3-1WH GAINFJT-57C1-1WH GAINFJT-58C1-1WH GAINFJT-58C3-4WH GAINFJT-62C1-0.5WH GAINFJT-62C3-1WH GAINFJT-67C3-1WH GAINFJT-7C1-1WH GAINFJT-7C3-5WH GAINFJT-7SBC1-1WH GAINFJT-85C1-2WH GAINFJT-97C1-1WH GAINFJT-97C3-2WH GAINFJT-98C1-1WH GAINFJT99C1-0.25WH GAINFJT-99C3-0.5WH GAINFJT-9C1-1WH GAINFJT-9C3-1WH GAINFJT-269C1-2WH
Rectangle Permanent Deep Freeze Labels for Thermal Transfer Printers
Etiquetas
These permanent labels are designed for the identification of tubes, vials, boxes, and other plastic or cardboard containers stored in ultra-low temperature laboratory freezers.
  • Temperature range: –80 to +110 °C (–112 to +230 °F)
  • Ideal for long-term storage in deep freeze conditions (–80 °C)
  • No cure time; can be stored in freezers immediately after application
  • No additional lamination necessary
  • Labels and printout resist spraying with alcohols and other chemicals
  • Printable with all major thermal-transfer printer brands
  • Available in a large range of sizes,