Deep-Freeze Removable Tapes
Proveedor: GA
TRM-13C1-50WH TRM-13C1-50BL TRM-13C1-50GA TRM-13C1-50OR TRM-13C1-50RE TRM-13C1-50YE TRM-13C3-100WH TRM-13C3-100BL TRM-13C3-100GA TRM-13C3-100OR TRM-13C3-100RE TRM-13C3-100YE TRM19SBC1-50WH TRM19SBC1-50BL TRM19SBC1-50GA TRM19SBC1-50OR TRM19SBC1-50RE TRM19SBC1-50YE TRM19SBC3100WH TRM19SBC3100BL TRM19SBC3100GA TRM19SBC3100OR TRM19SBC3100RE TRM19SBC3100YE TRM25SBC1-50WH TRM25SBC1-50BL TRM25SBC1-50GA TRM25SBC1-50OR TRM25SBC1-50RE TRM25SBC1-50YE TRM25SBC3100WH TRM25SBC3100BL TRM25SBC3100GA TRM25SBC3100OR TRM25SBC3100RE TRM25SBC3100YE
GAINTRM-13C1-50WHEA 27.7 EUR
GAINTRM-13C1-50WH GAINTRM-13C1-50BL GAINTRM-13C1-50GA GAINTRM-13C1-50OR GAINTRM-13C1-50RE GAINTRM-13C1-50YE GAINTRM-13C3-100WH GAINTRM-13C3-100BL GAINTRM-13C3-100GA GAINTRM-13C3-100OR GAINTRM-13C3-100RE GAINTRM-13C3-100YE GAINTRM19SBC1-50WH GAINTRM19SBC1-50BL GAINTRM19SBC1-50GA GAINTRM19SBC1-50OR GAINTRM19SBC1-50RE GAINTRM19SBC1-50YE GAINTRM19SBC3100WH GAINTRM19SBC3100BL GAINTRM19SBC3100GA GAINTRM19SBC3100OR GAINTRM19SBC3100RE GAINTRM19SBC3100YE GAINTRM25SBC1-50WH GAINTRM25SBC1-50BL GAINTRM25SBC1-50GA GAINTRM25SBC1-50OR GAINTRM25SBC1-50RE GAINTRM25SBC1-50YE GAINTRM25SBC3100WH GAINTRM25SBC3100BL GAINTRM25SBC3100GA GAINTRM25SBC3100OR GAINTRM25SBC3100RE GAINTRM25SBC3100YE
Deep-Freeze Removable Tapes
Etiquetas
Removable lab tape for deep-freeze storage of cryo containers, including freezer boxes, tubes, vials, plastic bags, and other items intended for long-term storage in laboratory freezers and dry ice transportation. Writable using permanent cryo markers.
  • Temperature range: −80 to 100 °C (−112 to 212 °F)
  • Resists long-term storage in deep-freeze conditions, including multiple freeze-thaw cycles
  • Glove-friendly and removable; won't leave behind any adhesive residue
  • Available in a range of colors for easy color coding
Deep-freeze removable tape for sealingMore Product Information