Transparent Cryogenic Tapes for Frozen Surfaces
Proveedor: GA
TXC-25CP3-100 TXC-19CP1-50 TXC-19CP3-100 TXC-25CP1-50
GAINTXC-25CP3-100EA 75.4 EUR
GAINTXC-25CP3-100 GAINTXC-19CP1-50 GAINTXC-19CP3-100 GAINTXC-25CP1-50
Transparent Cryogenic Tapes for Frozen Surfaces
Etiquetas
This clear liner-less cryogenic tape for already frozen surfaces is provided with innovative GatorCUT technology, making it effortlessly hand-cuttable. Perfect for identifying frozen samples and containers without concealing their contents, ensuring they remain visible.
  • Temperature range: –196 to +121 °C
  • Applicable on frozen surfaces, as low as –80 °C
  • Ideal for storage in ultra-low temperature conditions
  • Liner-free with GatorCUT technology; portable without the need for cutting instruments
  • Transparent design ensures contents of labeled containers remain