Anti-V5 Tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Genscript
A00877-40
GENSA00877-40EA 173 EUR
GENSA00877-40
Anti-V5 Tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Type: Tag
Antigen: V5 Tag (V5 tag)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR