Bis-Tris mini protein gels
Proveedor: Genscript
M41210 M41212 M41215 M42010 M42012 M42015 M00652 M00653 M00661 M00662 M00663 M00664 M00665 M00666 M00667 M00668 M00669 M00654 M00655 M00656 M00657 M00658 M00659 M00660
GENSM41210EA 263 EUR
GENSM41210 GENSM41212 GENSM41215 GENSM42010 GENSM42012 GENSM42015 GENSM00652 GENSM00653 GENSM00661 GENSM00662 GENSM00663 GENSM00664 GENSM00665 GENSM00666 GENSM00667 GENSM00668 GENSM00669 GENSM00654 GENSM00655 GENSM00656 GENSM00657 GENSM00658 GENSM00659 GENSM00660
Bis-Tris mini protein gels
Geles de electroforesis Geles prefabricados
GenScript's Bis-Tris gels are high-performing polyacrylamide gels designed to separate a wide range of protein sizes by electrophoresis. The gels are cast in a neutral pH buffer that minimises polyacrylamide hydrolysis and increases gel stability. Furthermore, the neutral pH which minimises protein modification when compared to Tris-glycine gels.
  • Large well volume
  • High resoluton and reproducibility
  • Long shelf life
  • Cost effective
  • Compatible cassette design
GenScript's SurePAGE™ Gels are premium, high resolution with enhance reproducibility gels, whileMore Product Information