Density hydrometers, M100
Proveedor: GERING
0345 0346 0347 0348 0349 0350 0351 0352 0344 0353 0354 0355
GERI0345EA 54.7 EUR
GERI0345 GERI0346 GERI0347 GERI0348 GERI0349 GERI0350 614-0115 614-0116 614-0193 GERI0353 GERI0354 GERI0355
Density hydrometers, M100
Hidrómetros
Reference temperature for density measurements: 20 °C.
Certificaciones: DIN 12791
Envase: In velvet lined case.