Tubing, Silicone, Sani-Tech®, ULTRA-C Series
Proveedor: Saint Gobain Life Sciences
ULTRA-C-012-0F ULTRA-C-020-0F ULTRA-C-025-0F ULTRA-C-030-0F ULTRA-C-030-2F ULTRA-C-040-0F ULTRA-C-058-0F ULTRA-C-062-1F ULTRA-C-062-2F ULTRA-C-062-3F ULTRA-C-062-4F ULTRA-C-062-5F ULTRA-C-078-1F ULTRA-C-078-2F ULTRA-C-078-3F ULTRA-C-078-5F ULTRA-C-093-1F ULTRA-C-093-2F ULTRA-C-093-3F ULTRA-C-093-4F ULTRA-C-125-1F ULTRA-C-125-2F ULTRA-C-125-3F ULTRA-C-125-4F ULTRA-C-125-5F ULTRA-C-156-4F ULTRA-C-156-5F ULTRA-C-187-1F ULTRA-C-187-2F ULTRA-C-187-3F ULTRA-C-187-4F ULTRA-C-187-5F ULTRA-C-187-6F ULTRA-C-250-1F ULTRA-C-250-2F ULTRA-C-250-3F ULTRA-C-250-4F ULTRA-C-312-1F ULTRA-C-312-2F ULTRA-C-312-3F ULTRA-C-312-4F ULTRA-C-312-5F ULTRA-C-375-1F ULTRA-C-375-2F ULTRA-C-375-3F ULTRA-C-375-4F ULTRA-C-375-5F ULTRA-C-375-6F ULTRA-C-500-1F ULTRA-C-500-2F ULTRA-C-500-3F ULTRA-C-500-4F ULTRA-C-500-5F ULTRA-C-500-6F ULTRA-C-625-1F ULTRA-C-625-2F ULTRA-C-625-3F ULTRA-C-625-4F ULTRA-C-625-5F ULTRA-C-625-6F ULTRA-C-750-1F ULTRA-C-750-2F ULTRA-C-750-3F ULTRA-C-750-4F ULTRA-C-750-5F ULTRA-C-750-6F ULTRA-C-875-1F ULTRA-C-875-2F ULTRA-C-875-4F ULTRA-C-875-5F
GESSULTRA-C-012-0FEA 90.2 EUR
GESSULTRA-C-012-0F GESSULTRA-C-020-0F GESSULTRA-C-025-0F GESSULTRA-C-030-0F GESSULTRA-C-030-2F GESSULTRA-C-040-0F GESSULTRA-C-058-0F GESSULTRA-C-062-1F GESSULTRA-C-062-2F GESSULTRA-C-062-3F GESSULTRA-C-062-4F GESSULTRA-C-062-5F GESSULTRA-C-078-1F GESSULTRA-C-078-2F GESSULTRA-C-078-3F GESSULTRA-C-078-5F GESSULTRA-C-093-1F GESSULTRA-C-093-2F GESSULTRA-C-093-3F GESSULTRA-C-093-4F GESSULTRA-C-125-1F GESSULTRA-C-125-2F GESSULTRA-C-125-3F GESSULTRA-C-125-4F GESSULTRA-C-125-5F GESSULTRA-C-156-4F GESSULTRA-C-156-5F GESSULTRA-C-187-1F GESSULTRA-C-187-2F GESSULTRA-C-187-3F GESSULTRA-C-187-4F GESSULTRA-C-187-5F GESSULTRA-C-187-6F GESSULTRA-C-250-1F GESSULTRA-C-250-2F GESSULTRA-C-250-3F GESSULTRA-C-250-4F GESSULTRA-C-312-1F GESSULTRA-C-312-2F GESSULTRA-C-312-3F GESSULTRA-C-312-4F GESSULTRA-C-312-5F GESSULTRA-C-375-1F GESSULTRA-C-375-2F GESSULTRA-C-375-3F GESSULTRA-C-375-4F GESSULTRA-C-375-5F GESSULTRA-C-375-6F GESSULTRA-C-500-1F GESSULTRA-C-500-2F GESSULTRA-C-500-3F GESSULTRA-C-500-4F GESSULTRA-C-500-5F GESSULTRA-C-500-6F GESSULTRA-C-625-1F GESSULTRA-C-625-2F GESSULTRA-C-625-3F GESSULTRA-C-625-4F GESSULTRA-C-625-5F GESSULTRA-C-625-6F GESSULTRA-C-750-1F GESSULTRA-C-750-2F GESSULTRA-C-750-3F GESSULTRA-C-750-4F GESSULTRA-C-750-5F GESSULTRA-C-750-6F GESSULTRA-C-875-1F GESSULTRA-C-875-2F GESSULTRA-C-875-4F GESSULTRA-C-875-5F
Tubing, Silicone, Sani-Tech®, ULTRA-C Series
Tuberías Tubo de transferencia
Sani-Tech® ultra-pure platinum-cured silicone tubing meets the stringent requirements of the biopharmaceutical market.
  • Maximum temperature range: –62 to +260 °C (–80 to +500 °F)
  • Imparts no taste or odour to critical stream
  • Available in 50 and 65 durometers
  • Laser etched for lot traceability
It features ultra low extractables, excellent flexibility, longer pump life and non-yellowing formulation.
Applications include sterile fill and transfers, drug delivery, cell harvest, media processing, sanitary peristaltic pump tubing, production filtration andMore Product Information